Learn More
Cole-Parmer pH electrode, general purpose, combination, glass body, Delrin shield, BNC
Catalog No. 50633507 Shop All Cole-Parmer Products
Encompass_Preferred
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
Catalog No. 50-633-507 Supplier Cole-Parmer Supplier No. 0599710
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.