Learn More
Cole-Parmer Oakton by Cole-Parmer pH electrode, combination, spear tip, sealed, glass body, BNC
Catalog No. 50633513 Shop All Cole-Parmer Products
Encompass_Preferred
Glass-body combination electrodes feature refillable references, except for 05998-30 which features a high-temperature KNO3 gel. All have BNC connectors.
Catalog No. 50-633-513 Supplier Cole-Parmer Supplier No. 0599820
Glass-body combination electrodes feature refillable references, except for 05998-30 which features a high-temperature KNO3 gel. All have BNC connectors.