Learn More
Cole-Parmer Cole-Parmer pH electrode, Refillable/SJ/Glass/Extra-Long/Thin/326x8mm; BNC
Catalog No. 50633511 Shop All Cole-Parmer Products
Encompass_Preferred
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
Catalog No. 50-633-511 Supplier Cole-Parmer Supplier No. 0599040
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.