Learn More
Cole-Parmer Cole-Parmer Tuff-Tip pH Electrode, 3/4" (100 Ohms RTD), 3.5mm phono plug
Supplier:  Cole-Parmer 2702042
Encompass_Preferred
Don't worry about bulb breakage, the unique Tuff-Tip design helps protect bulbs. Choose chemical-resistant Ryton PPS standard electrode or with graphite tip. Eliminate electrical noise with graphite-tipped models. Ideal for viscous or severe applications, these electrodes reduce contamination and clogging with uniform pore size.
Catalog No. 50-633-701