Learn More
Cole-Parmer Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC
Catalog No. 50633472 Shop All Cole-Parmer Products
Encompass_Preferred
An electrode with a semi-domed tuff-tip limits glass exposure and protects from damage. For your in-line process control needs, it is available in a single or double junction and BNC or tinned ends.
Catalog No. 50-633-472 Supplier Cole-Parmer Supplier No. 0599514
An electrode with a semi-domed tuff-tip limits glass exposure and protects from damage. For your in-line process control needs, it is available in a single or double junction and BNC or tinned ends.