Learn More
Cole-Parmer Cole-Parmer Insertion/Submersion pH Probe; DJ/CPVC/; /BNC
Supplier:  Cole-Parmer 2730120
Encompass_Preferred
In-line or submersible electrodes with probe guard operate from 23 to 176°>F (-5 to 80°>C) and have a PVC housing with a 3/4'' NPT(M) fitting on both ends. Epoxy body withstands pressures to 100 psi. Measure pH from 0 to 14. All electrodes measure 6-3/4''L x 1'' OD, with a 1-3/4'' insertion depth.
Catalog No. 50-633-451