Learn More
Cole-Parmer Cole-Parmer In-Line pH Electrode with Tuff-Tip, No ATC Element
Catalog No. 50633465 Shop All Cole-Parmer Products
Encompass_Preferred
An electrode with a semi-domed tuff-tip limits glass exposure and protects from damage. For your in-line process control needs, it is available in a single or double junction and BNC or tinned ends.
Catalog No. 50-633-465 Supplier Cole-Parmer Supplier No. 0599501
An electrode with a semi-domed tuff-tip limits glass exposure and protects from damage. For your in-line process control needs, it is available in a single or double junction and BNC or tinned ends.