Learn More
Cole-Parmer Oakton by Cole-Parmer In-Line pH Probe, SJ/PVDF/100Ohm RTD/10'; Plug/BNC
Catalog No. 50633462 Shop All Cole-Parmer Products
Encompass_Preferred
In-line electrodes operate from 32 to 212°F (0 to 100°C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 4'' x 15/16'' diameter. Electrode extends 1'' beyond housing. Maximum pressure 100 psi. Measure pH from 0 to 13.
Catalog No. 50-633-462 Supplier Cole-Parmer Supplier No. 0599370
In-line electrodes operate from 32 to 212°F (0 to 100°C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 4'' x 15/16'' diameter. Electrode extends 1'' beyond housing. Maximum pressure 100 psi. Measure pH from 0 to 13.