Learn More
Cole-Parmer Cole-Parmer Insertion/Submersion pH Probe; DJ/CPVC/100Ohm RTD; Plug/BNC
Catalog No. 50633452 Shop All Cole-Parmer Products
Encompass_Preferred
These in-line or submersible electrodes with probe guard operate from 23 to 176 degrees Fahrenheit or -5 to 80 degrees Celsius. They have a PVC housing with a 3/4 inch NPT(M) fitting on both ends. It's epoxy body withstands pressures up to 100 psi, and measures pH from 0 to 14. All electrodes measure 6-3/4 inches long by one inch outer diameter with a 1-3/4 inch insertion depth.
Catalog No. 50-633-452 Supplier Cole-Parmer Supplier No. 2730121
These in-line or submersible electrodes with probe guard operate from 23 to 176 degrees Fahrenheit or -5 to 80 degrees Celsius. They have a PVC housing with a 3/4 inch NPT(M) fitting on both ends. It's epoxy body withstands pressures up to 100 psi, and measures pH from 0 to 14. All electrodes measure 6-3/4 inches long by one inch outer diameter with a 1-3/4 inch insertion depth.