Learn More
Cole-Parmer Cole-Parmer Insertion/Submersion pH Probe; SJ/CPVC/100Ohm RTD; Plug/BNC
Catalog No. 50633448 Shop All Cole-Parmer Products
Encompass_Preferred
In-line or submersible electrodes with probe guard operate from 23 to 176>F (-5 to 80>C) and have a PVC housing with a 3/4'' NPT(M) fitting on both ends. Epoxy body withstands pressures to 100 psi. Measure pH from 0 to 14. All electrodes measure 6-3/4''L x 1'' OD, with a 1-3/4'' insertion depth.
Catalog No. 50-633-448 Supplier Cole-Parmer Supplier No. 2730021
In-line or submersible electrodes with probe guard operate from 23 to 176>F (-5 to 80>C) and have a PVC housing with a 3/4'' NPT(M) fitting on both ends. Epoxy body withstands pressures to 100 psi. Measure pH from 0 to 14. All electrodes measure 6-3/4''L x 1'' OD, with a 1-3/4'' insertion depth.