Learn More
Cole-Parmer Oakton by Cole-Parmer pH electrode, in-line, single-junction, no ATC, 10-ft cable, BNC
Catalog No. 50633461 Shop All Cole-Parmer Products
Encompass_Preferred
This is an integral, one piece electrode and housing for installation convenience. Choose from single-junction or double-junction designs. It is available with BNC or tinned ends.
Catalog No. 50-633-461 Supplier Cole-Parmer Supplier No. 0599390
This is an integral, one piece electrode and housing for installation convenience. Choose from single-junction or double-junction designs. It is available with BNC or tinned ends.