Learn More
Cole-Parmer Oakton Single-junction, semi-dome, polygel reference, temperature sensor w/ waterproof connection (Anthony) pH electrode, 100C, 12 mm x 115mm,30K ATC.
Catalog No. 501961379 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1379 Supplier Cole-Parmer Supplier No. 3580888
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.