Learn More
Cole-Parmer Cole-Parmer Solution Grounded pH Probe, DJ/PPS/No ATC; BNC
Catalog No. 50633646 Shop All Cole-Parmer Products
Encompass_Preferred
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton housing with 3/4'' NPT(M) threads.
Catalog No. 50-633-646 Supplier Cole-Parmer Supplier No. 2701800
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton housing with 3/4'' NPT(M) threads.