Learn More
Cole-Parmer Cole-Parmer Solution Grounded pH Probe, DJ/PPS/100Ohm RTD; Tinned/BNC
Catalog No. 50633648 Shop All Cole-Parmer Products
Encompass_Preferred
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton housing with 3/4'' NPT(M) threads.
Catalog No. 50-633-648 Supplier Cole-Parmer Supplier No. 2701803
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton housing with 3/4'' NPT(M) threads.