Learn More
Cole-Parmer Cole-Parmer Solution Grounded pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
Catalog No. 50633647 Shop All Cole-Parmer Products
Encompass_Preferred
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-foot cable with BNC pH connector and Ryton housing with 3/4 inch NPT(M) threads.
Catalog No. 50-633-647 Supplier Cole-Parmer Supplier No. 2701802
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-foot cable with BNC pH connector and Ryton housing with 3/4 inch NPT(M) threads.