Learn More
Cole-Parmer Oakton by Cole-Parmer Autoclavable pH Probe, Sealed/DJ/Glass; 210 mm
Catalog No. 50633695 Shop All Cole-Parmer Products
Encompass_Preferred
These electrodes are designed for insertion in RADIOMETER mountings or Ingold nonpressurized assemblies 761-351, 761-361, and 361CH-361 (12 mm dia.) Sealed, gel-filled, double-junction reference electrodes require no refilling, and offer long-term stability and extend service life.
Catalog No. 50-633-695 Supplier Cole-Parmer Supplier No. 0599396
These electrodes are designed for insertion in RADIOMETER mountings or Ingold nonpressurized assemblies 761-351, 761-361, and 361CH-361 (12 mm dia.) Sealed, gel-filled, double-junction reference electrodes require no refilling, and offer long-term stability and extend service life.