Learn More
Cole-Parmer Oakton double-junction pH electrode, standard bulb, polygel reference, temperature sensor w/ waterproof connection, 100°C, 12mm x 110mm, 30K
Catalog No. 501961380 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1380 Supplier Cole-Parmer Supplier No. 3580889
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.