Learn More
Cole-Parmer Cole-Parmer pH Electrodes for Field Use; single-junction 10 foot cable
Catalog No. 50633525 Shop All Cole-Parmer Products
Encompass_Preferred
These combination electrodes withstand the most demanding field conditions—the semi-domed tip protects the pH bulb and ensures a quick response when taking measurements in harsh and dirty environments. Easy-to-clean electrodes feature an impact-resistant epoxy body for further protection from the elements. The large, annular ceramic reference junction and sealed cross-linked polymer reference resist clogging. Operating range is from 0 to 14 pH. Electrodes measure 110 mm L x 12 mm dia and withstand temperatures up to 212°F (100°C). All models have BNC connectors.
Catalog No. 50-633-525 Supplier Cole-Parmer Supplier No. 0599204
These combination electrodes withstand the most demanding field conditions—the semi-domed tip protects the pH bulb and ensures a quick response when taking measurements in harsh and dirty environments. Easy-to-clean electrodes feature an impact-resistant epoxy body for further protection from the elements. The large, annular ceramic reference junction and sealed cross-linked polymer reference resist clogging. Operating range is from 0 to 14 pH. Electrodes measure 110 mm L x 12 mm dia and withstand temperatures up to 212°F (100°C). All models have BNC connectors.