Learn More
Cole-Parmer Cole-Parmer Electrode pH Soil Glass BNC
Supplier:  Cole-Parmer 0599262
Encompass_Preferred
Our refillable soil electrode has an extra-wide 7.5 mm ceramic junction. Soil electrodes have a maximum temperature range of 212°F (100°C) and measure 145 mm L x 12 mm dia. Ag/AgCl ref cell. Use with any meter that accepts a BNC connector, or check out our selection of pH/Ion meters.
Catalog No. 50-633-562