Learn More
Cole-Parmer Oakton by Cole-Parmer Submersible 3' L pH Probe, SJ/ABS; BNC
Catalog No. 50633494 Shop All Cole-Parmer Products
Encompass_Preferred
This submersible electrode operates from 32 to 176>F (0 to 80>C). Submerse it up to the cable and measure pH from 0 to 13.
Catalog No. 50-633-494 Supplier Cole-Parmer Supplier No. 0599380
This submersible electrode operates from 32 to 176>F (0 to 80>C). Submerse it up to the cable and measure pH from 0 to 13.