Learn More
Cole-Parmer Cole-Parmer Electrode pH submersible 50 ft cable length BNC
Catalog No. 501961676 Shop All Cole-Parmer Products
Encompass_Preferred
These ABS plastic electrodes are designed for field applications. Annular junction provides fast response and resists pressure effects of submersion. BNC connection is compatible with most pH meters. 100 foot cable length electrode includes preamplification.
Catalog No. 50-196-1676 Supplier Cole-Parmer Supplier No. 3580554
These ABS plastic electrodes are designed for field applications. Annular junction provides fast response and resists pressure effects of submersion. BNC connection is compatible with most pH meters. 100 foot cable length electrode includes preamplification.