Learn More
Cole-Parmer Oakton by Cole-Parmer Submersible 3' L pH Probe, DJ/ABS/100Ohm RTD; Plug/BNC
Catalog No. 50633498 Shop All Cole-Parmer Products
Encompass_Preferred
Submersible electrodes operate from 32 to 176>F (0 to 80>C) and can submerse up to the cable. Electrodes are potted in a 3 foot length by 1-1/16 inch diameter ABS sheath. These electrodes are ideal for measurements in open containers, such as tanks or troughs.
Catalog No. 50-633-498 Supplier Cole-Parmer Supplier No. 2700181
Submersible electrodes operate from 32 to 176>F (0 to 80>C) and can submerse up to the cable. Electrodes are potted in a 3 foot length by 1-1/16 inch diameter ABS sheath. These electrodes are ideal for measurements in open containers, such as tanks or troughs.