Learn More
Cole-Parmer Cole-Parmer Submersible 3' L pH Probe, SJ/ABS/100Ohm RTD; Tinned/BNC
Catalog No. 50633496 Shop All Cole-Parmer Products
Encompass_Preferred
Submersible electrodes operate from 32 to 176°>F (0 to 80°>C). Submerse up to the cable. Measure pH from 0 to 13. Electrodes are potted in a 3 ft L x 1-1/16'' diameter ABS sheath. Ideal for measurements in open containers such as tanks or troughs.
Catalog No. 50-633-496 Supplier Cole-Parmer Supplier No. 0599385
Submersible electrodes operate from 32 to 176°>F (0 to 80°>C). Submerse up to the cable. Measure pH from 0 to 13. Electrodes are potted in a 3 ft L x 1-1/16'' diameter ABS sheath. Ideal for measurements in open containers such as tanks or troughs.