Learn More
Cole-Parmer Oakton by Cole-Parmer Submersible 3' L pH Probe, SJ/ABS/100Ohm RTD; Plug/BNC
Catalog No. 50633495 Shop All Cole-Parmer Products
Encompass_Preferred
This submersible electrode operates from 32 to 176>F (0 to 80>C). Submerse it up to the cable and measure pH from 0 to 13.
Catalog No. 50-633-495 Supplier Cole-Parmer Supplier No. 0599381
This submersible electrode operates from 32 to 176>F (0 to 80>C). Submerse it up to the cable and measure pH from 0 to 13.