Learn More
Cole-Parmer Cole-Parmer All-in-One pH/ATC Probe, Refillable/DJ/Glass
Catalog No. 50633575 Shop All Cole-Parmer Products
Encompass_Preferred
Use these electrodes with most Oakton handheld or benchtop pH meters for accurate and reliable readings. They are refillable and provide a more accurate pH measurement. An amber glass bulb minimizes sodium error in high pH solutions. Use the electrode at temperatures up to 100 degrees Celsius.
Catalog No. 50-633-575 Supplier Cole-Parmer Supplier No. 0599136
Use these electrodes with most Oakton handheld or benchtop pH meters for accurate and reliable readings. They are refillable and provide a more accurate pH measurement. An amber glass bulb minimizes sodium error in high pH solutions. Use the electrode at temperatures up to 100 degrees Celsius.