Learn More
Cole-Parmer Oakton "All-in-one" pH Probe with ATC for 300/310 Series Meters
Catalog No. 50633523 Shop All Cole-Parmer Products
Encompass_Preferred
This replacement all-in-one pH / temperature single-junction electrode is non-refillable. 12-mm dia. BNC pH connection, 6 Pin ATC connection for Oakton waterproof meters.
Catalog No. 50-633-523 Supplier Cole-Parmer Supplier No. 5900275
This replacement all-in-one pH / temperature single-junction electrode is non-refillable. 12-mm dia. BNC pH connection, 6 Pin ATC connection for Oakton waterproof meters.