Learn More
Cole-Parmer Cole-Parmer pH Probe, DJ/Glass/100Ohm RTD/12 mm/10'; Plug/BNC
Catalog No. 50633570 Shop All Cole-Parmer Products
Encompass_Preferred
This general purpose, double-junction electrode prevents contamination of the AgCl reference cell by solutions that might react with silver, such as heavy metals, organics and sulfides.
Catalog No. 50-633-570 Supplier Cole-Parmer Supplier No. 0599427
This general purpose, double-junction electrode prevents contamination of the AgCl reference cell by solutions that might react with silver, such as heavy metals, organics and sulfides.