Learn More
Cole-Parmer Cole-Parmer pH Probe, DJ/Glass/1kOhm RTD/12 mm/10'; Spade/BNC
Catalog No. 50633571 Shop All Cole-Parmer Products
Encompass_Preferred
Use this probe for general purpose, double-junction electrodes to prevent contamination of the AgCl reference cell by solutions that might react with silver, such as heavy metals, organics and sulfides.
Catalog No. 50-633-571 Supplier Cole-Parmer Supplier No. 0565482
Use this probe for general purpose, double-junction electrodes to prevent contamination of the AgCl reference cell by solutions that might react with silver, such as heavy metals, organics and sulfides.