Learn More
Cole-Parmer Cole-Parmer Removable High pH Double-Junction pH Electrode
Catalog No. 50633491 Shop All Cole-Parmer Products
Encompass_Preferred
These electrodes are ideal for applications involving high-sodium concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Use for measurements from 0 to 14 pH. Electrodes withstand temperatures from 23 to 230ºF (-5 to 110ºC).
Catalog No. 50-633-491 Supplier Cole-Parmer Supplier No. 2700920
These electrodes are ideal for applications involving high-sodium concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Use for measurements from 0 to 14 pH. Electrodes withstand temperatures from 23 to 230ºF (-5 to 110ºC).