Learn More
Cole-Parmer Oakton by Cole-Parmer Ion-Selective Electrode (ISE), Potassium, Double-Junction, BNC
Catalog No. 50633658 Shop All Cole-Parmer Products
Encompass_Preferred
Easily fit this electrode of potassium with glass body into small spaces without any added reference electrode.
Catalog No. 50-633-658 Supplier Cole-Parmer Supplier No. 2750239
Easily fit this electrode of potassium with glass body into small spaces without any added reference electrode.