Learn More
AB Sciex LLC Electrode Assembly Replacement For use only with CESI 8000 PA 800 plus or PA 800 Enhanced instruments
Catalog No. NC1038122
Encompass
The electrode has a PEEK sleeve adding strength and rigidity. It features a snap-in design for easy removal and replacement. The platinum electrode is for use only with CESI 8000 PA 800 plus or PA 800 Enhanced instruments.
Catalog No. NC1038122 Supplier AB Sciex LLC Supplier No. A47775
The electrode has a PEEK sleeve adding strength and rigidity. It features a snap-in design for easy removal and replacement. The platinum electrode is for use only with CESI 8000 PA 800 plus or PA 800 Enhanced instruments.