Learn More
Cole-Parmer Tuff-Tip, Semi-Dome 1" In-Line/Sub pH, graphite, 1kOhm spade lug ATC
Catalog No. NC1644143 Shop All Cole-Parmer Products
Encompass_Preferred
Don't worry about bulb breakage— unique design helps protect bulbs – Eliminate electrical noise with graphite-tipped models – Ideal for viscous or severe applications – Reduce contamination and clogging with uniform pore size
Catalog No. NC1644143 Supplier Cole-Parmer Supplier No. 2702066
Don't worry about bulb breakage— unique design helps protect bulbs – Eliminate electrical noise with graphite-tipped models – Ideal for viscous or severe applications – Reduce contamination and clogging with uniform pore size