Learn More
Cole-Parmer Cole-Parmer Gel-Filled Side-Mount pH Probe, DJ/PVDF/100hm RTD; Tinned/BNC
Catalog No. 50633480 Shop All Cole-Parmer Products
Encompass_Preferred
Sealed AgCl reference cell requires no refilling during use. Rugged PVDF housing with 1/2'' NPT(M) thread withstands pressures up 100psi, good for measurements up to 14 pH units. Includes a 10-ft cable with BNC connector.
Catalog No. 50-633-480 Supplier Cole-Parmer Supplier No. 2701130
Sealed AgCl reference cell requires no refilling during use. Rugged PVDF housing with 1/2'' NPT(M) thread withstands pressures up 100psi, good for measurements up to 14 pH units. Includes a 10-ft cable with BNC connector.