Learn More
Electron Microscopy Sciences Electrode Single Hole Flex Arm
SDP
Catalog No. 5029266
Encompass_Preferred
This flexible arm assists by holding and positioning electrodes or other objects in an direction. The base is solid polyethylene and comes complete with 3-M double sided mounting tape on the bottom to secure the unit to the table top. The unit is available in either a one or two hole head configuration.
Catalog No. 50-292-66 Supplier Electron Microscopy Sciences Supplier No. 6436601
This flexible arm assists by holding and positioning electrodes or other objects in an direction. The base is solid polyethylene and comes complete with 3-M double sided mounting tape on the bottom to secure the unit to the table top. The unit is available in either a one or two hole head configuration.