Learn More
Cole-Parmer Oakton by Cole-Parmer Ion-Selective Electrode (ISE), Sodium, Double-Junction, BNC
Catalog No. 50633667 Shop All Cole-Parmer Products
Encompass_Preferred
Easily fit this electrode of sodium with glass body into small spaces without any added reference electrode.
Catalog No. 50-633-667 Supplier Cole-Parmer Supplier No. 2750243
Easily fit this electrode of sodium with glass body into small spaces without any added reference electrode.