Learn More
Cole-Parmer Oakton by Cole-Parmer Combination Ion-Selective Electrode (ISE), Sodium (Na+)
Catalog No. 50633668 Shop All Cole-Parmer Products
Encompass_Preferred
Easily fit this electrode of sodium with epoxy body into small spaces without any added reference electrode.
Catalog No. 50-633-668 Supplier Cole-Parmer Supplier No. 2750430
Easily fit this electrode of sodium with epoxy body into small spaces without any added reference electrode.