Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Electrode Storage Solution
SCI_ED

Catalog No. S25907 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25907 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25907
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For storing pH electrodes.

  • Poly bottle packaging
  • Volume: 1L

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Type Solution
Filling Solution Type Storage
For Use With (Application) Storing pH electrodes
Packaging Poly Bottle
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.