Learn More
Sensorex ELECTRODE STORAGE SOLUTION
Catalog No. NC9973750
Encompass_Preferred
Sensorex general use electrode storage solution. Keeps any pH electrode wet and instantly ready Continuously soaks pH and ORP electrodes in solution to prevent electrode dry out For use with most pH / ORP electrodes in the market Highest Quality Solutions 1pint
Catalog No. NC9973750 Supplier Sensorex Supplier No. S016
Sensorex general use electrode storage solution. Keeps any pH electrode wet and instantly ready Continuously soaks pH and ORP electrodes in solution to prevent electrode dry out For use with most pH / ORP electrodes in the market Highest Quality Solutions 1pint