Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electrode Storage Solution for Storing Glass and Combination pH Electrodes, Reagents

Catalog No. p-200021836
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-403 1 L
01-335-401 4 L
01-335-400 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-335-403 Supplier Reagents Holdings Llc Supplier No. CE0016001A
Only null left
Add to Cart
Add to Cart

Electrode Storage Solution for Storing Glass and Combination pH Electrodes, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 100°C
Color Colorless
For Use With (Equipment) Electrode
Packaging Poly Bottle
Quantity 1 L
Solubility Completely miscible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.