Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Electrode Support
SDPSCI_ED

Catalog No. S16042ND Shop All Vernier Software Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S16042ND Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16042ND Supplier Vernier™ Supplier No. ESUP
Add to Cart
Add to Cart

Includes

Electrode Support

Our Electrode Support is a great complement to the Vernier Stir Station, as well as a perfect holder for many sensors.

It is built to connect to all standard ring-stand posts; its large-handled locking nut keeps your sensors firmly in place. The larger opening (13 mm in diameter) is perfect for our pH, ISE, Conductivity and ORP sensors. The smaller opening (5 mm) is designed to hold our Stainless Steel Temperature Probe or the Vernier Go!Temp.

Warranty and Services

5-year limited warranty

Specifications

Color Black
For Use With (Equipment) Electrode
Type Support Clamp
For Use With (Application) Educational use only
Diameter (Metric) Small Hole 5 mm, Large Hole 13 mm
Diameter (Metric) Cap 0.5 to 5 cm Clamp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.