Learn More
Med Vet International ECG Resting Electrode Welch Allyn Paper Backing Tab Connector, 108071, 100/Pack
Catalog No. 503044095
Encompass_Preferred
Sold by the case, 5000 electrodes per case Synthetic paper material Conductive Adhesive Gel Patented traction liner prevents tabs from sliding off liner saving time and money Pliable substrate conforms to body Ag/Ag chloride
Catalog No. 50-304-4095 Supplier Med Vet International Supplier No. 1166165PK
Sold by the case, 5000 electrodes per case Synthetic paper material Conductive Adhesive Gel Patented traction liner prevents tabs from sliding off liner saving time and money Pliable substrate conforms to body Ag/Ag chloride