Learn More
Med Vet International Ecg Monitoring Electrode Foam Backing Radiolucent / Mr Conditional Snap Connector, 2570, 1000/Case
Catalog No. 502832079
Encompass_Preferred
Built in -purpose monitoring electrodes with Foam Backing backing and sticky gel are ideally suited for the OR, ER and diagnostic stress testing applications Conveniently packaged in individual strips of ten electrodes for easy use and less waste Remains fresh for 30 days out of the bag High performance adhesive offers instant adhesion and up to five-day wear with excellent patient skin care and comfort Sticky gel delivers excellent trace quality resulting in better patient Trace Prep surface gently prepares skin for optimal trace quality Radiolucent
Catalog No. 50-283-2079 Supplier Med Vet International Supplier No. 368361CS
Built in -purpose monitoring electrodes with Foam Backing backing and sticky gel are ideally suited for the OR, ER and diagnostic stress testing applications Conveniently packaged in individual strips of ten electrodes for easy use and less waste Remains fresh for 30 days out of the bag High performance adhesive offers instant adhesion and up to five-day wear with excellent patient skin care and comfort Sticky gel delivers excellent trace quality resulting in better patient Trace Prep surface gently prepares skin for optimal trace quality Radiolucent