Learn More
Crescent Chemical Co Inc ELECTRODE WICKS EXTRA SIZE
SDP
Catalog No. 50248723
Encompass_Preferred
Electrode wicks extra size, 300 x 6 x 1 mm Filter cardboard, 300 x 6 x 1 mm. Size - 100PIECES Storage Conditions - +15 °C TO +30 °C Catalog Number - 42972.03
Catalog No. 50-248-723 Supplier Crescent Chemical Co Inc Supplier No. SE42972.03
Electrode wicks extra size, 300 x 6 x 1 mm Filter cardboard, 300 x 6 x 1 mm. Size - 100PIECES Storage Conditions - +15 °C TO +30 °C Catalog Number - 42972.03