Learn More
Hach Company Electrodeless Cond. Sens PVDF
Catalog No. 5000208037 Shop All Hach Company Products
Encompass
Digital Electrodeless Conductivity Sensor-Convertible Body Style-PVDF Body Material-Digital Gateway-1m Extension CablePVDF Convertible Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable
Catalog No. 50-002-08037 Supplier Hach Company Supplier No. D3726E2T
Digital Electrodeless Conductivity Sensor-Convertible Body Style-PVDF Body Material-Digital Gateway-1m Extension CablePVDF Convertible Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable