Learn More
Hach Company Electrodeless Conduc. Sens PFA
Catalog No. 5000207776 Shop All Hach Company Products
Encompass
Digital Electrodeless Conductivity Sensor-Sanitary Body Style-PFA Body MaterialPFA Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable
Catalog No. 50-002-07776 Supplier Hach Company Supplier No. D3708E2T
Digital Electrodeless Conductivity Sensor-Sanitary Body Style-PFA Body MaterialPFA Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable