Learn More
Hach Company Electrodeless Conduct Sens-San
Catalog No. 5000207644 Shop All Hach Company Products
Encompass
Electrodeless Conductivity Sensor-Sanitary Style-Flange-PP Body Material-50-ft CablePolypropylene Sanitary (CIP) Flange Mounting Style 15m (50ft) Analog Cable Electrodeless Conductivity Sensor
Catalog No. 50-002-07644 Supplier Hach Company Supplier No. 3705E2T50N
Electrodeless Conductivity Sensor-Sanitary Style-Flange-PP Body Material-50-ft CablePolypropylene Sanitary (CIP) Flange Mounting Style 15m (50ft) Analog Cable Electrodeless Conductivity Sensor