Learn More
Hach Company Electrodeless Conduct Sens-San
Catalog No. 5000207693 Shop All Hach Company Products
Encompass
Sanitary Electrodeless Conductivity Sensor-PFA Body MaterialPFA Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Generic ID Electrodeless Conductivity Sensor
Catalog No. 50-002-07693 Supplier Hach Company Supplier No. 3708E2T20G
Sanitary Electrodeless Conductivity Sensor-PFA Body MaterialPFA Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Generic ID Electrodeless Conductivity Sensor