Learn More
Hach Company Electrodeless (inductive) conductivity sensor - sanitary (CIP) flange, PFA body, 6 m (20 ft) analog cable, built-in Pt1000 RTD
Catalog No. 5000207645 Shop All Hach Company Products
Encompass
This electrodeless inductive conductivity sensor is designed for sanitary process environments and clean-in-place (CIP) applications. It features a chemically resistant PFA body and is intended for reliable online conductivity monitoring, membrane protection, and process control where wide-range measurement and temperature compensation are required.
  • PFA body for chemical resistance and durability.
  • Sanitary flange mounting for CIP compatibility.
  • Electrodeless design for stable measurement across a wide range.
  • Built-in Pt1000 RTD for accurate temperature compensation.
  • 6 m analog cable for flexible installation options.
  • Measurement range suitable for low to very high conductivities.
  • Includes sensor, cable, and user manual for quick setup.
Catalog No. 50-002-07645 Supplier Hach Company Supplier No. 5000TC007000
This electrodeless inductive conductivity sensor is designed for sanitary process environments and clean-in-place (CIP) applications. It features a chemically resistant PFA body and is intended for reliable online conductivity monitoring, membrane protection, and process control where wide-range measurement and temperature compensation are required.
  • PFA body for chemical resistance and durability.
  • Sanitary flange mounting for CIP compatibility.
  • Electrodeless design for stable measurement across a wide range.
  • Built-in Pt1000 RTD for accurate temperature compensation.
  • 6 m analog cable for flexible installation options.
  • Measurement range suitable for low to very high conductivities.
  • Includes sensor, cable, and user manual for quick setup.