Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Electroless nickel plating solution

Catalog No. 9016551L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-165-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-165-51L Supplier Sigma Aldrich Supplier No. 9016551L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Stable electroless nickel plating composition, ready to use, specified for making ohmic contacts to silicon and other semiconductor materials.
  • Advantages.
  • Stable, ready to use.
  • Plates without generating ammonia fumes.
  • Produces quality electrical contacts on p- and n-type silicon.
  • Does not compensate for p- or n-type conductivity.
  • Excellent adhesion and solderability.
  • Plates on silicon, germanium, gallium arsenide, and cadmium sulfide
  • Nickelex is a vastly improved, electroless nickel preparation, formulated specially for semiconductor use. The composition is based upon ions of a nickel complex and hypophosphite together in solution with a catalytic accelerator and a stabilizer. The solution is also buffered at the optimum pH for electroless nickel catalysis. In addition, only high-purity chemical materials with trace impurities (such as copper) specifically removed are utilized. NICKELEX is a very stable product, free of ammonia, and ready for use without the need for any addition or mixing. Fume problems are eliminated. Nickelex, furthermore, is not subject to undesirable changes in composition during use. The chemical reaction during the electroless plating process involves an oxidation-reduction reaction between nickelous and hypophosphite ions.
  • The reaction is essentially a two-step process occurring simultaneously.
  • The reaction progresses catalytically with nickel deposition occurring at the operating temperature of 95°C to 100°C. The nascent hydrogen ensures an oxide-free nickel deposit. In addition, some NiP formed in a side reaction improves corrosion resistance and solderability. The nickel deposit is highly conductive, remains unstressed, particularly after suitable heat treatment, and shows good adherence.
  • Application
  • Use of Nickelex:
  • Nickelex provides good mechanical and electrical contacts for semiconductor devices (silicon rectifiers, solar cells, varactors, microwave diodes, transistors, microcircuits, etc.). The quality of the electrical contact is excellent if the surface of silicon is abraded by lapping to increase surface recombination velocity; if the surface is highly doped from diffusion; or if the semiconductor resistivity is low (0.1 ohm-cm). Nickelex also produces adherent deposits on germanium, gallium arsenide, and cadmium sulfide.
  • The Nickelex plate exhibits excellent solderability with lead, tin, and Pb/Sn alloys. Flux can be omitted when the furnace is soldered in a hydrogen or non-oxidizing atmosphere.
  • Instructions
  • Complete metallization procedures are provided for silicon rectifier and planar device fabrication. These procedures should be followed to obtain optimum adhesion of nickel and quality of the ohmic contacts.

Specifications

Quantity 1 L
Synonym Nickelex
Biological Activity Lungs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.