Learn More
Metrohm USA ELECTROLYTE 3 ML KCL
SDP
Catalog No. NC9894061 Shop All Metrohm USA Products
Encompass
Electrolyte 3 mol/L KCl; 250 mL. For ag/agCl reference systems.
Catalog No. NC9894061 Supplier Metrohm USA Supplier No. 62308020
Electrolyte 3 mol/L KCl; 250 mL. For ag/agCl reference systems.